Assay Details
| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133906.4 | NP_598667.2 | 4 - 5 | 897 | 62 | |
| GenBank mRNA | AK020039.1 | - | 4 - 5 | 873 | 62 | |
| AK028932.1 | - | 4 - 5 | 828 | 62 | ||
| AK031759.1 | - | 4 - 5 | 878 | 62 | ||
| AK036807.1 | - | 4 - 5 | 880 | 62 | ||
| AK054323.1 | - | 4 - 5 | 868 | 62 | ||
| AK135999.1 | - | 4 - 5 | 847 | 62 | ||
| AK154831.1 | - | 4 - 5 | 877 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zkscan1
Genomic Map