Assay Details
| Gene Symbol: Fam83h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168253.1 | NP_001161725.1 | 3 - 4 | 757 | 62 | |
| NM_134087.2 | NP_598848.2 | 3 - 4 | 725 | 62 | ||
| GenBank mRNA | AK030392.1 | - | 3 - 4 | 759 | 62 | |
| AK035531.1 | - | 3 - 4 | 725 | 62 | ||
| BC117947.1 | - | 3 - 4 | 698 | 40098049 | 62 | |
| BC117948.1 | - | 3 - 4 | 698 | 40098059 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam83h
Genomic Map