Assay Details
| Gene Symbol: Gstm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010359.2 | NP_034489.1 | 1 - 2 | 70 | 59 | |
| GenBank mRNA | BC044927.1 | - | 1 - 2 | 54 | 5051815 | 59 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gstm3
Genomic Map