Assay Details
| Gene Symbol: Gm35438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045464.1 | - | 3 - 4 | 864 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 815 | Gm815 | -285.4kb | Anti-sense | Chr. 19 - 26885925 - 26888647 on NCBI Build GRCm38 | |
| Downstream | very low density lipoprotein receptor | Vldlr |
AA408956, AI451093, AW047288 |
41.7kb | Anti-sense | Chr. 19 - 27216489 - 27254231 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm35438
Genomic Map