Assay Details
| Gene Symbol: Mlec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175403.3 | NP_780612.2 | 2 - 3 | 551 | 67 | |
| GenBank mRNA | AK031525.1 | - | 2 - 3 | 521 | 67 | |
| AK075940.1 | - | 2 - 3 | 535 | 67 | ||
| AK129066.1 | - | 2 - 3 | 454 | 67 | ||
| BC035300.1 | - | 2 - 3 | 516 | 4237864 | 67 | |
| BC058245.1 | - | 2 - 3 | 505 | 6313259 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlec
Genomic Map