Assay Details
| Gene Symbol: Mlec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175403.3 | NP_780612.2 | 3 - 4 | 728 | 137 | |
| GenBank mRNA | AK031525.1 | - | 3 - 4 | 698 | 137 | |
| AK075940.1 | - | 3 - 4 | 712 | 137 | ||
| AK129066.1 | - | 3 - 4 | 631 | 137 | ||
| BC035300.1 | - | 3 - 4 | 693 | 4237864 | 137 | |
| BC058245.1 | - | 3 - 4 | 682 | 6313259 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlec
Genomic Map