Assay Details
| Gene Symbol: Ankrd13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026718.2 | NP_080994.2 | 10 - 11 | 1161 | 71 | |
| GenBank mRNA | AK039432.1 | - | 10 - 11 | 1356 | 71 | |
| AK156539.1 | - | 10 - 11 | 1354 | 71 | ||
| AK162340.1 | - | 2 - 3 | 182 | 71 | ||
| BC027765.1 | - | 8 - 9 | 836 | 71 | ||
| BC049187.1 | - | 10 - 11 | 1340 | 71 | ||
| BC061692.1 | - | 10 - 11 | 1161 | 6308996 | 71 | |
| BC108413.1 | - | 10 - 11 | 1636 | 6487251 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd13a
Genomic Map