Assay Details
| Gene Symbol: Ankrd13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026718.2 | NP_080994.2 | 12 - 13 | 1433 | 89 | |
| GenBank mRNA | AK039432.1 | - | 12 - 13 | 1628 | 89 | |
| AK156539.1 | - | 12 - 13 | 1626 | 89 | ||
| AK162340.1 | - | 4 - 5 | 454 | 89 | ||
| BC003286.1 | - | 2 - 3 | 160 | 89 | ||
| BC027765.1 | - | 10 - 11 | 1108 | 89 | ||
| BC049187.1 | - | 12 - 13 | 1612 | 89 | ||
| BC061692.1 | - | 12 - 13 | 1433 | 6308996 | 89 | |
| BC108413.1 | - | 12 - 13 | 1908 | 6487251 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd13a
Genomic Map