Assay Details
| Gene Symbol: 4930429D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040699.1 | 5 - 6 | 486 | 67 | ||
| GenBank mRNA | AK015233.1 | - | 5 - 6 | 487 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase 10 | Mapk10 |
C230008H04Rik, JNK3, JNK3B1, JNK3B2, SAPK(beta), Serk2, p493F12, p54bSAPK |
-29.4kb | Sense | Chr. 5 - 102907948 - 103211339 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700021F02 gene | 1700021F02Rik | 92.1kb | Anti-sense | Chr. 5 - 103346535 - 103348302 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930429D17Rik
Genomic Map