Assay Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028041.2 | NP_082317.1 | 5 - 6 | 624 | 95 | |
| GenBank mRNA | AF319547.1 | - | 5 - 6 | 984 | 95 | |
| AK010484.1 | - | 2 - 3 | 204 | 95 | ||
| AK028274.1 | - | 5 - 6 | 638 | 95 | ||
| AK031534.1 | - | 5 - 6 | 634 | 95 | ||
| AK147715.1 | - | 5 - 6 | 639 | 95 | ||
| AK152989.1 | - | 5 - 6 | 638 | 95 | ||
| AK155138.1 | - | 5 - 6 | 636 | 95 | ||
| AK167603.1 | - | 5 - 6 | 638 | 95 | ||
| AK168455.1 | - | 5 - 6 | 638 | 95 | ||
| AK169255.1 | - | 5 - 6 | 637 | 95 | ||
| AK170967.1 | - | 5 - 6 | 636 | 95 | ||
| AK212480.1 | - | 1 - 2 | 51 | 95 | ||
| BC043699.1 | - | 5 - 6 | 624 | 5344158 | 95 | |
| BC055026.1 | - | 1 - 2 | 67 | 95 | ||
| BC062920.1 | - | 5 - 6 | 627 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx54
Genomic Map