Assay Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028041.2 | NP_082317.1 | 10 - 11 | 1076 | 66 | |
| GenBank mRNA | AF319547.1 | - | 10 - 11 | 1436 | 66 | |
| AK010484.1 | - | 7 - 8 | 656 | 66 | ||
| AK028274.1 | - | 10 - 11 | 1090 | 66 | ||
| AK031534.1 | - | 10 - 11 | 1086 | 66 | ||
| AK031679.1 | - | 10 - 11 | 1094 | 66 | ||
| AK147715.1 | - | 10 - 11 | 1091 | 66 | ||
| AK155138.1 | - | 10 - 11 | 1088 | 66 | ||
| AK167603.1 | - | 10 - 11 | 1090 | 66 | ||
| AK168455.1 | - | 10 - 11 | 1090 | 66 | ||
| AK169255.1 | - | 10 - 11 | 1089 | 66 | ||
| AK170967.1 | - | 10 - 11 | 1088 | 66 | ||
| BC021900.1 | - | 1 - 2 | 81 | 66 | ||
| BC043699.1 | - | 10 - 11 | 1076 | 5344158 | 66 | |
| BC055026.1 | - | 6 - 7 | 519 | 66 | ||
| BC062920.1 | - | 10 - 11 | 1079 | 66 | ||
| BC071251.1 | - | 4 - 5 | 373 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx54
Genomic Map