Assay Details
| Gene Symbol: Gatc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029645.3 | NP_083921.1 | 2 - 3 | 425 | 87 | |
| GenBank mRNA | AK008080.1 | - | 2 - 3 | 430 | 87 | |
| AK034326.1 | - | 2 - 3 | 425 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24009120 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24009122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24009123 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24009121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24009124 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P174661 | AK034326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P174661 | NM_029645.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P174661 | AK008080.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P174661 | AK034326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P174661 | NM_029645.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P174661 | AK008080.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P424272 | AK047064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P424272 | BC021775.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P424272 | AK034326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P424272 | NM_029645.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P424272 | AK047064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P424272 | BC021775.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P424272 | AK034326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P424272 | NM_029645.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944360 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336702 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848722 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569446 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84810 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179340 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P424272 | AK047064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P424272 | BC021775.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P424272 | AK034326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P424272 | NM_029645.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P424272 | AK047064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P424272 | BC021775.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P424272 | AK034326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P424272 | NM_029645.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P174661 | AK034326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P174661 | NM_029645.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P174661 | AK008080.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P174661 | AK034326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P174661 | NM_029645.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P174661 | AK008080.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174661 | AK034326.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174661 | NM_029645.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174661 | AK008080.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174661 | AK034326.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174661 | NM_029645.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174661 | AK008080.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24009121 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179340 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05706117 | - | - |
| Whole Mouse Genome G4122A | A_52_P174661 | AK034326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P174661 | NM_029645.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P174661 | AK008080.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P174661 | AK034326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P174661 | NM_029645.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P174661 | AK008080.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P424272 | AK047064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P424272 | BC021775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P424272 | AK034326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P424272 | NM_029645.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P424272 | AK047064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P424272 | BC021775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P424272 | AK034326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P424272 | NM_029645.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129996 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179340 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04980778 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04534957 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04534956 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04980776 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24009124 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24009123 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24009121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24009120 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24009122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24009111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04215187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04258052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04288716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04215179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04551171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04603567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04980778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24009101 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10532991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532992 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532990 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425211_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447568_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10532989 | - | - |
| Mouse Genome 430 2.0 Array | 1425211_at | - | - |
| Mouse Genome 430 2.0 Array | 1447568_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447568_at | - | - |
| Mouse Genome 430 2.0 Array | 1425211_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 384281_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 384281_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500027578.mm.1 | TC0500003096.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500027578.mm.1 | NM_029645.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500027578.mm.1 | NM_029645.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500027579.mm.1 | TC0500003096.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500027579.mm.1 | NM_029645.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500027579.mm.1 | NM_029645.3 | Refseq |
| Mouse Expression Set 430 | 1425211_PM_at | - | - |
| Mouse Expression Set 430 | 1425211_at | - | - |
| Mouse Expression Set 430 | 1447568_at | - | - |
| Mouse Expression Set 430 | 1447568_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425211_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425211_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425211_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425211_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447568_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447568_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 835183 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100520605 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100520605 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734941 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236821 | - | - |
| mouse-6 v1.1 (Illumina) | scl0384281.2_10-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100520605 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100520605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2807665 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2807665 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2734941 | - | - |

Gatc
Genomic Map