Assay Details
| Gene Symbol: D930016D06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040297.1 | - | 1 - 2 | 738 | 109 | |
| BC082597.1 | - | 1036 | 109 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC005561 | BC005561 | -18.1kb | Sense | Chr. 5 - 104508352 - 104522383 on NCBI Build GRCm38 | |
| Downstream | zinc finger protein 33B | Zfp33b | 2310012P17Rik | 21.2kb | Sense | Chr. 5 - 104570873 - 104584620 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

D930016D06Rik
Genomic Map