Assay Details
| Gene Symbol: Prmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133740.2 | NP_598501.1 | 14 - 15 | 1512 | 103 | |
| GenBank mRNA | AK010534.1 | - | 14 - 15 | 1523 | 103 | |
| AK031738.1 | - | 14 - 15 | 1513 | 103 | ||
| AK086646.1 | - | 14 - 15 | 1471 | 103 | ||
| AK155236.1 | - | 14 - 15 | 1510 | 103 | ||
| BC008128.1 | - | 10 - 11 | 1055 | 103 | ||
| BC050775.1 | - | 14 - 15 | 1503 | 6534023 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt3
Genomic Map