Assay Details
| Gene Symbol: Prmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133740.2 | NP_598501.1 | 4 - 5 | 410 | 153 | |
| GenBank mRNA | AK008118.1 | - | 4 - 5 | 402 | 153 | |
| AK010534.1 | - | 4 - 5 | 422 | 153 | ||
| AK031738.1 | - | 4 - 5 | 411 | 153 | ||
| AK049836.1 | - | 4 - 5 | 391 | 153 | ||
| AK086646.1 | - | 4 - 5 | 369 | 153 | ||
| AK155236.1 | - | 4 - 5 | 408 | 153 | ||
| BC050775.1 | - | 4 - 5 | 389 | 6534023 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt3
Genomic Map