Assay Details
| Gene Symbol: Prmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133740.2 | NP_598501.1 | 6 - 7 | 668 | 71 | |
| GenBank mRNA | AK010534.1 | - | 6 - 7 | 680 | 71 | |
| AK031738.1 | - | 6 - 7 | 669 | 71 | ||
| AK086646.1 | - | 6 - 7 | 627 | 71 | ||
| AK155236.1 | - | 6 - 7 | 666 | 71 | ||
| AK164064.1 | - | 1 - 2 | 2019 | 71 | ||
| BC008128.1 | - | 2 - 3 | 211 | 71 | ||
| BC050775.1 | - | 6 - 7 | 647 | 6534023 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt3
Genomic Map