Assay Details
| Gene Symbol: Nell1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037906.2 | NP_001032995.1 | 3 - 4 | 372 | 129 | |
| GenBank mRNA | AY622226.1 | - | 3 - 4 | 372 | 129 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK035921.1 | - | 861 | Chr. - - [-] on Build | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nell1
Genomic Map