Assay Details
| Gene Symbol: Scaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008422.1 | NP_001008422.1 | 3 - 4 | 347 | 85 | |
| GenBank mRNA | AK141920.1 | - | 4 - 5 | 361 | 85 | |
| BC042576.1 | - | 3 - 4 | 205 | 4160122 | 85 | |
| BC043138.1 | - | 3 - 4 | 377 | 6821344 | 85 | |
| BC054818.1 | - | 2 - 3 | 557 | 85 | ||
| BC079873.1 | - | 3 - 4 | 365 | 85 | ||
| BC085153.1 | - | 3 - 4 | 347 | 6837651 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scaf1
Genomic Map