Assay Details
| Gene Symbol: Scaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008422.1 | NP_001008422.1 | 4 - 5 | 439 | 84 | |
| GenBank mRNA | AK141920.1 | - | 5 - 6 | 453 | 84 | |
| BC042576.1 | - | 4 - 5 | 297 | 4160122 | 84 | |
| BC043138.1 | - | 4 - 5 | 469 | 6821344 | 84 | |
| BC054818.1 | - | 3 - 4 | 649 | 84 | ||
| BC079873.1 | - | 4 - 5 | 457 | 84 | ||
| BC085153.1 | - | 4 - 5 | 439 | 6837651 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scaf1
Genomic Map