Assay Details
| Gene Symbol: Gm19303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045092.1 | 1 - 2 | 87 | |||
| GenBank mRNA | AK046060.1 | - | 1 - 2 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | trichorhinophalangeal syndrome I (human) | Trps1 |
AI115454, AI447310, D15Ertd586e |
-455.0kb | Sense | Chr. 15 - 50654757 - 50890747 on NCBI Build GRCm38 |
| Downstream |
eukaryotic translation initiation factor 3, subunit H |
Eif3h |
1110008A16Rik, 40kD, 9430017H16Rik, EIF3-P40, EIF3-gamma, Eif3s3 |
391.0kb | Sense | Chr. 15 - 51786560 - 51865515 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm19303
Genomic Map