Assay Details
| Gene Symbol: Gm19744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032387.1 | - | 1 - 2 | 3179 | 150 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | teneurin transmembrane protein 3 | Tenm3 |
2610100B16Rik, Odz1, Odz3, Ten-m3, mKIAA1455 |
Anti-sense | Chr. 8 - 48225665 - 48935162 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 1700019L22 gene | 1700019L22Rik | 236.9kb | Anti-sense | Chr. 8 - 49096147 - 50918746 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm19744
Genomic Map