Assay Details
Gene Symbol: 1700019L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006131.1 | - | 3 - 4 | 371 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 2516 | Gm2516 | Anti-sense | Chr. 8 - 50368112 - 50416089 on NCBI Build GRCm38 | ||
Downstream |
predicted gene, 32975 |
Gm32975 | 762.5kb | Anti-sense | Chr. 8 - 51177683 - 51180608 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205183 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868419 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973876 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894006 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124490 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803266 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322475 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813785 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499393 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374285 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973028 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241768 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923784 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759481 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744516 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875134 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754489 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309738 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465907 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573660 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770424 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374146 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491074 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644376 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839673 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368895 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731891 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219702 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204166 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525569 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172304 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226864 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508796 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710676 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768190 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341500 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139449 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46734 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166923 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123727 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35990 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 21477 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168250 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06509858 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06509882 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06509797 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06509837 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06509762 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06509959 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06509911 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125459 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06509837 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06509959 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06509882 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860340 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06788659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06751700 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860699 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860628 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860754 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06928203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860174 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860574 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860598 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860592 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860702 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860329 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860487 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07085367 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860218 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06928204 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860709 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07271382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860760 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860480 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06872912 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860691 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860632 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06928201 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06748852 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06702782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860601 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860240 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860325 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860607 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06748858 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860494 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860619 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07432376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860615 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860327 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860694 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06866523 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860741 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07339979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860337 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07411902 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860230 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860493 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06751701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06669010 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860497 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860486 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06723535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860491 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860596 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860485 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860612 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860724 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860611 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860335 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860618 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860333 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860642 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860717 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860225 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860603 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07142247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860489 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07276662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860681 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860332 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860609 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07005720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860625 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860495 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860762 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07411920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860630 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860470 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25860787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06702783 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Murine Genome U74v2 Set | 171264_at | - | - |
Murine Genome U74v2 Set | 171264_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106760338 | - | - |
