Assay Details
| Gene Symbol: Lipn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027340.2 | NP_081616.1 | 5 - 6 | 764 | 126 | |
| GenBank mRNA | AK008974.1 | - | 4 - 5 | 394 | 126 | |
| AK154333.1 | - | 5 - 6 | 765 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lipn
Genomic Map