Assay Details
| Gene Symbol: C730002L08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045778.1 | 1 - 2 | 139 | 77 | ||
| GenBank mRNA | AK050017.1 | - | 1 - 2 | 140 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

C730002L08Rik
Genomic Map