Assay Details
| Gene Symbol: Vwce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027913.1 | NP_082189.1 | 1 - 2 | 353 | 70 | |
| GenBank mRNA | AK005010.1 | - | 1 - 2 | 354 | 70 | |
| AK153937.1 | - | 1 - 2 | 133 | 70 | ||
| BC030181.1 | - | 1 - 2 | 305 | 70 | ||
| BC043473.1 | - | 1 - 2 | 293 | 5346280 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vwce
Genomic Map