Assay Details
| Gene Symbol: Tead2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285498.1 | NP_001272427.1 | 1 - 2 | 126 | 108 | |
| GenBank mRNA | Y10026.1 | - | 1 - 2 | 48 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001285498.1, Y10026.1 transcripts.
Set Membership: |
|

Tead2
Genomic Map