Assay Details
| Gene Symbol: Pold1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011131.3 | NP_035261.3 | 11 - 12 | 1437 | 62 | |
| GenBank mRNA | AK088310.1 | - | 11 - 12 | 1428 | 62 | |
| AK146837.1 | - | 11 - 12 | 1434 | 62 | ||
| AK166592.1 | - | 11 - 12 | 1631 | 62 | ||
| AK167569.1 | - | 11 - 12 | 1420 | 62 | ||
| AK168967.1 | - | 11 - 12 | 1400 | 62 | ||
| AK169040.1 | - | 11 - 12 | 1433 | 62 | ||
| BC009128.1 | - | 11 - 12 | 1438 | 3257174 | 62 | |
| BC052670.1 | - | 10 - 11 | 1404 | 30058314 | 62 | |
| Z21848.1 | - | 11 - 12 | 1406 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pold1
Genomic Map