Assay Details
| Gene Symbol: Gm15787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040431.1 | 2 - 3 | 268 | 66 | ||
| GenBank mRNA | AK053084.1 | - | 2 - 3 | 269 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 605 | Zfp605 | A830023I12Rik | -37.0kb | Anti-sense | Chr. 5 - 110110022 - 110130509 on NCBI Build GRCm38 |
| Downstream |
golgi autoantigen, golgin subfamily a, 3 |
Golga3 |
5330413L04, 5430416E01Rik, AI449376, AW490576, G1-499-14, Mea-2, Mea2, repro27 |
6.3kb | Anti-sense | Chr. 5 - 110176659 - 110226470 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm15787
Genomic Map