Assay Details
| Gene Symbol: A430057O09 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079755.1 | - | 2 - 3 | 305 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein kinase, cGMP-dependent, type II |
Prkg2 |
AW212535, CGKII, Prkgr2, cGK-II |
-129.6kb | Sense | Chr. 5 - 98929773 - 99038718 on NCBI Build GRCm38 |
| Downstream |
RasGEF domain family, member 1B |
Rasgef1b |
4732452O09Rik, GPIgamma4, Gpig4 |
47.0kb | Sense | Chr. 5 - 99217420 - 99252927 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

A430057O09
Genomic Map