Assay Details
| Gene Symbol: A430057O09 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079755.1 | - | 4 - 5 | 674 | 186 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein kinase, cGMP-dependent, type II |
Prkg2 |
AW212535, CGKII, Prkgr2, cGK-II |
-80.1kb | Sense | Chr. 5 - 98929773 - 99038718 on NCBI Build GRCm38 |
| Downstream |
RasGEF domain family, member 1B |
Rasgef1b |
4732452O09Rik, GPIgamma4, Gpig4 |
93.7kb | Sense | Chr. 5 - 99217420 - 99252927 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

A430057O09
Genomic Map