Assay Details
| Gene Symbol: Gm10390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045793.1 | 2 - 3 | 687 | 81 | ||
| GenBank mRNA | AK041808.1 | - | 2 - 3 | 686 | 81 | |
| AK169507.1 | - | 2 - 3 | 686 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 38554 |
Gm38554 | -157.7kb | Anti-sense | Chr. 5 - 119959177 - 119963380 on NCBI Build GRCm38 | |
| Downstream | LIM homeobox protein 5 | Lhx5 | Lim2 | 306.2kb | Anti-sense | Chr. 5 - 120431886 - 120441457 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK169507.1 transcripts.
Set Membership: |
|

Gm10390
Genomic Map