Assay Details
| Gene Symbol: Bcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243052.1 | NP_001229981.1 | 5 - 6 | 787 | 76 | |
| NM_001243053.1 | NP_001229982.1 | 6 - 7 | 672 | 76 | ||
| NM_009737.3 | NP_033867.1 | 5 - 6 | 760 | 76 | ||
| GenBank mRNA | AF031467.1 | - | 5 - 6 | 552 | 76 | |
| AK037482.1 | - | 5 - 6 | 565 | 76 | ||
| AK137049.1 | - | 6 - 7 | 723 | 76 | ||
| AK145300.1 | - | 5 - 6 | 605 | 76 | ||
| AK160159.1 | - | 5 - 6 | 788 | 76 | ||
| BC017688.1 | - | 5 - 6 | 537 | 4242224 | 76 | |
| BC048072.1 | - | 5 - 6 | 551 | 4166264 | 76 | |
| U68526.1 | - | 5 - 6 | 546 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bcat2
Genomic Map