Assay Details
| Gene Symbol: Bcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243052.1 | NP_001229981.1 | 7 - 8 | 1097 | 75 | |
| NM_001243053.1 | NP_001229982.1 | 8 - 9 | 982 | 75 | ||
| NM_009737.3 | NP_033867.1 | 7 - 8 | 1070 | 75 | ||
| GenBank mRNA | AF031467.1 | - | 7 - 8 | 862 | 75 | |
| AK137049.1 | - | 8 - 9 | 1033 | 75 | ||
| AK145300.1 | - | 7 - 8 | 915 | 75 | ||
| AK160159.1 | - | 7 - 8 | 1098 | 75 | ||
| BC017688.1 | - | 7 - 8 | 847 | 4242224 | 75 | |
| BC048072.1 | - | 7 - 8 | 861 | 4166264 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcat2
Genomic Map