Assay Details
| Gene Symbol: 4930414N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040294.1 | 1 - 2 | 80 | 111 | ||
| GenBank mRNA | AK015139.1 | - | 1 - 2 | 80 | 111 | |
| AK076690.1 | - | 1 - 2 | 80 | 111 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | paired box 2 | Pax2 |
Opdc, Pax-2 |
-135.1kb | Anti-sense | Chr. 19 - 44745695 - 44837871 on NCBI Build GRCm38 |
| Downstream |
sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4G |
Sema4g |
AI507908, AW554132 |
4.4kb | Anti-sense | Chr. 19 - 44988966 - 45003397 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930414N06Rik
Genomic Map