Assay Details
| Gene Symbol: 6530437J22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020101.1 | - | 2 - 3 | 476 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis-specific serine kinase substrate | Tsks |
Stk22s1, Tssks1 |
-15.0kb | Sense | Chr. 7 - 44929577 - 44958035 on NCBI Build GRCm38 |
| Downstream | protein arginine N-methyltransferase 1 | Prmt1 |
6720434D09Rik, AW214366, Hrmt1l2, Mrmt1 |
3.0kb | Anti-sense | Chr. 7 - 44976755 - 44986420 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

6530437J22Rik
Genomic Map