Assay Details
| Gene Symbol: 6530437J22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020101.1 | - | 3 - 4 | 548 | 80 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DV064059.1 | - | 234 | Chr. - - [-] on Build | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis-specific serine kinase substrate | Tsks |
Stk22s1, Tssks1 |
-15.7kb | Sense | Chr. 7 - 44929577 - 44958035 on NCBI Build GRCm38 |
| Downstream | protein arginine N-methyltransferase 1 | Prmt1 |
6720434D09Rik, AW214366, Hrmt1l2, Mrmt1 |
2.5kb | Anti-sense | Chr. 7 - 44976755 - 44986420 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

6530437J22Rik
Genomic Map