Assay Details
| Gene Symbol: Rab35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198163.1 | NP_937806.1 | 2 - 3 | 265 | 80 | |
| GenBank mRNA | AB232631.1 | - | 2 - 3 | 105 | 80 | |
| AK156576.1 | - | 2 - 3 | 256 | 80 | ||
| BC056466.1 | - | 2 - 3 | 265 | 5717246 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab35
Genomic Map