Assay Details
| Gene Symbol: Tjp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198985.1 | NP_001185914.1 | 17 - 18 | 2708 | 100 | |
| NM_011597.4 | NP_035727.2 | 17 - 18 | 2698 | 100 | ||
| GenBank mRNA | AF113005.1 | - | 17 - 18 | 2556 | 100 | |
| AK028494.1 | - | 17 - 18 | 2625 | 100 | ||
| BC034677.1 | - | 17 - 18 | 2616 | 3155078 | 100 | |
| BC039924.1 | - | 17 - 18 | 2627 | 3155079 | 100 | |
| BC042695.1 | - | 17 - 18 | 2600 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tjp2
Genomic Map