Assay Details
| Gene Symbol: Tjp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198985.1 | NP_001185914.1 | 6 - 7 | 1194 | 86 | |
| NM_011597.4 | NP_035727.2 | 6 - 7 | 1184 | 86 | ||
| GenBank mRNA | AF113005.1 | - | 6 - 7 | 1042 | 86 | |
| AK028494.1 | - | 6 - 7 | 1113 | 86 | ||
| AK217702.1 | - | 1 - 2 | 96 | 86 | ||
| BC034677.1 | - | 6 - 7 | 1102 | 3155078 | 86 | |
| BC039924.1 | - | 6 - 7 | 1113 | 3155079 | 86 | |
| BC042695.1 | - | 6 - 7 | 1086 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tjp2
Genomic Map