Assay Details
| Gene Symbol: Tjp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198985.1 | NP_001185914.1 | 7 - 8 | 1344 | 68 | |
| NM_011597.4 | NP_035727.2 | 7 - 8 | 1334 | 68 | ||
| GenBank mRNA | AK028494.1 | - | 7 - 8 | 1263 | 68 | |
| BC034677.1 | - | 7 - 8 | 1252 | 3155078 | 68 | |
| BC039924.1 | - | 7 - 8 | 1263 | 3155079 | 68 | |
| BC042695.1 | - | 7 - 8 | 1236 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tjp2
Genomic Map