Assay Details
| Gene Symbol: Tjp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198985.1 | NP_001185914.1 | 8 - 9 | 1452 | 78 | |
| NM_011597.4 | NP_035727.2 | 8 - 9 | 1442 | 78 | ||
| GenBank mRNA | AK028494.1 | - | 8 - 9 | 1371 | 78 | |
| AK194560.1 | - | 1 - 2 | 84 | 78 | ||
| AK204133.1 | - | 1 - 2 | 84 | 78 | ||
| BC034677.1 | - | 8 - 9 | 1360 | 3155078 | 78 | |
| BC039924.1 | - | 8 - 9 | 1371 | 3155079 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tjp2
Genomic Map