Assay Details
| Gene Symbol: Plod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011962.3 | NP_036092.1 | 14 - 15 | 2287 | 96 | |
| GenBank mRNA | AF046783.1 | - | 14 - 15 | 1695 | 96 | |
| AK013195.1 | - | 14 - 15 | 1833 | 96 | ||
| AK033360.1 | - | 14 - 15 | 1832 | 96 | ||
| AK088948.1 | - | 14 - 15 | 1704 | 96 | ||
| AK157741.1 | - | 13 - 14 | 1906 | 96 | ||
| AK180845.1 | - | 1 - 2 | 72 | 96 | ||
| AK217183.1 | - | 1 - 2 | 72 | 96 | ||
| BC043047.1 | - | 14 - 15 | 2286 | 5703967 | 96 | |
| BC054734.1 | - | 14 - 15 | 2287 | 5720686 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plod3
Genomic Map