Assay Details
| Gene Symbol: Plod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011962.3 | NP_036092.1 | 3 - 4 | 1014 | 128 | |
| GenBank mRNA | AF046783.1 | - | 3 - 4 | 422 | 128 | |
| AK013195.1 | - | 3 - 4 | 560 | 128 | ||
| AK033360.1 | - | 3 - 4 | 559 | 128 | ||
| AK053320.1 | - | 3 - 4 | 656 | 128 | ||
| AK088948.1 | - | 3 - 4 | 431 | 128 | ||
| AK157741.1 | - | 3 - 4 | 557 | 128 | ||
| BC043047.1 | - | 3 - 4 | 1013 | 5703967 | 128 | |
| BC054734.1 | - | 3 - 4 | 1014 | 5720686 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plod3
Genomic Map