Assay Details
| Gene Symbol: Plod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011962.3 | NP_036092.1 | 6 - 7 | 1353 | 79 | |
| GenBank mRNA | AF046783.1 | - | 6 - 7 | 761 | 79 | |
| AK013195.1 | - | 6 - 7 | 899 | 79 | ||
| AK033360.1 | - | 6 - 7 | 898 | 79 | ||
| AK053320.1 | - | 6 - 7 | 995 | 79 | ||
| AK088948.1 | - | 6 - 7 | 770 | 79 | ||
| AK157741.1 | - | 6 - 7 | 862 | 79 | ||
| BC043047.1 | - | 6 - 7 | 1352 | 5703967 | 79 | |
| BC054734.1 | - | 6 - 7 | 1353 | 5720686 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plod3
Genomic Map