Assay Details
| Gene Symbol: Mtnr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008639.2 | NP_032665.1 | 1 - 2 | 189 | 63 | |
| GenBank mRNA | BC116900.1 | - | 1 - 2 | 262 | 40126219 | 63 |
| BC119313.1 | - | 1 - 2 | 262 | 8734062 | 63 | |
| U52222.1 | - | 1 - 2 | 189 | 63 | ||
More Information
Set Membership: |
|

Mtnr1a
Genomic Map