Assay Details
| Gene Symbol: Rad21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009009.4 | NP_033035.3 | 4 - 5 | 615 | 153 | |
| GenBank mRNA | AF332085.1 | - | 3 - 4 | 370 | 153 | |
| AF332086.1 | - | 3 - 4 | 370 | 153 | ||
| AK128984.1 | - | 4 - 5 | 504 | 153 | ||
| AK139492.1 | - | 3 - 4 | 268 | 153 | ||
| AK145591.1 | - | 4 - 5 | 582 | 153 | ||
| AK163992.1 | - | 476 | 153 | |||
| AK168718.1 | - | 4 - 5 | 603 | 153 | ||
| AK168896.1 | - | 4 - 5 | 564 | 153 | ||
| BC043032.1 | - | 3 - 4 | 384 | 5696540 | 153 | |
| BC129916.1 | - | 3 - 4 | 414 | 40111820 | 153 | |
| BC129917.1 | - | 3 - 4 | 415 | 40111821 | 153 | |
| D37790.1 | - | 3 - 4 | 302 | 153 | ||
| X98293.1 | - | 3 - 4 | 370 | 153 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad21
Genomic Map