Assay Details
| Gene Symbol: Rad21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009009.4 | NP_033035.3 | 7 - 8 | 1062 | 76 | |
| GenBank mRNA | AF332085.1 | - | 6 - 7 | 817 | 76 | |
| AF332086.1 | - | 6 - 7 | 817 | 76 | ||
| AK004746.1 | - | 7 - 8 | 865 | 76 | ||
| AK128984.1 | - | 7 - 8 | 951 | 76 | ||
| AK129050.1 | - | 2 - 3 | 190 | 76 | ||
| AK139492.1 | - | 6 - 7 | 715 | 76 | ||
| AK145591.1 | - | 7 - 8 | 1029 | 76 | ||
| AK163992.1 | - | 923 | 76 | |||
| AK168718.1 | - | 7 - 8 | 1050 | 76 | ||
| AK168896.1 | - | 7 - 8 | 1011 | 76 | ||
| BC043032.1 | - | 6 - 7 | 831 | 5696540 | 76 | |
| BC129916.1 | - | 6 - 7 | 861 | 40111820 | 76 | |
| BC129917.1 | - | 6 - 7 | 862 | 40111821 | 76 | |
| D37790.1 | - | 6 - 7 | 749 | 76 | ||
| X98293.1 | - | 6 - 7 | 817 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad21
Genomic Map