Assay Details
| Gene Symbol: Rad21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009009.4 | NP_033035.3 | 9 - 10 | 1405 | 112 | |
| GenBank mRNA | AF332085.1 | - | 8 - 9 | 1160 | 112 | |
| AF332086.1 | - | 8 - 9 | 1160 | 112 | ||
| AK004746.1 | - | 9 - 10 | 1208 | 112 | ||
| AK128984.1 | - | 9 - 10 | 1294 | 112 | ||
| AK129050.1 | - | 4 - 5 | 533 | 112 | ||
| AK139492.1 | - | 8 - 9 | 1058 | 112 | ||
| AK145591.1 | - | 9 - 10 | 1372 | 112 | ||
| AK163992.1 | - | 1266 | 112 | |||
| AK168718.1 | - | 9 - 10 | 1393 | 112 | ||
| AK168896.1 | - | 9 - 10 | 1354 | 112 | ||
| BC043032.1 | - | 8 - 9 | 1174 | 5696540 | 112 | |
| BC129916.1 | - | 8 - 9 | 1204 | 40111820 | 112 | |
| BC129917.1 | - | 8 - 9 | 1205 | 40111821 | 112 | |
| X98293.1 | - | 8 - 9 | 1160 | 112 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rad21
Genomic Map