Assay Details
| Gene Symbol: Eif3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080635.1 | NP_542366.1 | 4 - 5 | 562 | 92 | |
| GenBank mRNA | AK088436.1 | - | 4 - 5 | 585 | 92 | |
| AK146200.1 | - | 4 - 5 | 585 | 92 | ||
| AK149081.1 | - | 4 - 5 | 574 | 92 | ||
| AK159113.1 | - | 4 - 5 | 575 | 92 | ||
| AK160335.1 | - | 5 - 6 | 586 | 92 | ||
| AK167651.1 | - | 4 - 5 | 565 | 92 | ||
| AK167815.1 | - | 4 - 5 | 583 | 92 | ||
| AK167844.1 | - | 4 - 5 | 585 | 92 | ||
| AK167928.1 | - | 4 - 5 | 589 | 92 | ||
| AK168108.1 | - | 4 - 5 | 587 | 92 | ||
| AK168123.1 | - | 4 - 5 | 605 | 92 | ||
| AK168259.1 | - | 4 - 5 | 585 | 92 | ||
| AK168274.1 | - | 4 - 5 | 585 | 92 | ||
| AK168553.1 | - | 4 - 5 | 587 | 92 | ||
| AK169416.1 | - | 4 - 5 | 586 | 92 | ||
| BC014755.1 | - | 4 - 5 | 562 | 4504013 | 92 | |
| BC086915.1 | - | 4 - 5 | 584 | 30122747 | 92 | |
| BC091728.1 | - | 4 - 5 | 570 | 2647034 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif3h
Genomic Map