Assay Details
| Gene Symbol: Kctd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159941.1 | NP_001153413.1 | 1 - 2 | 79 | 77 | |
| NM_026145.4 | NP_080421.2 | 1 - 2 | 79 | 77 | ||
| GenBank mRNA | AK028657.1 | - | 1 - 2 | 46 | 77 | |
| AK041516.1 | - | 1 - 2 | 46 | 77 | ||
| AK145295.1 | - | 1 - 2 | 58 | 77 | ||
| AK166505.1 | - | 1 - 2 | 58 | 77 | ||
| BB855690.1 | - | 1 - 2 | 79 | 77 | ||
| BC006935.1 | - | 1 - 2 | 60 | 3601917 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028657.1, AK041516.1, AK145295.1, AK166505.1, BC006935.1, NM_001159941.1, NM_026145.4 transcripts.
Set Membership: |
|

Kctd10
Genomic Map